Assay Details
| Gene Symbol: Lamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017959.2 | NP_001017959.1 | 6 - 7 | 1035 | 122 | |
| NM_001290485.1 | NP_001277414.1 | 6 - 7 | 1035 | 122 | ||
| NM_010685.4 | NP_034815.2 | 6 - 7 | 1035 | 122 | ||
| GenBank mRNA | AK012149.1 | - | 6 - 7 | 994 | 122 | |
| AK032974.1 | - | 6 - 7 | 997 | 122 | ||
| AK048174.1 | - | 6 - 7 | 977 | 122 | ||
| AK078193.1 | - | 6 - 7 | 1029 | 122 | ||
| AK088804.1 | - | 7 - 8 | 1012 | 122 | ||
| AK128946.1 | - | 4 - 5 | 697 | 122 | ||
| AK136550.1 | - | 6 - 7 | 996 | 122 | ||
| AK146731.1 | - | 6 - 7 | 1016 | 122 | ||
| AK150766.1 | - | 6 - 7 | 1004 | 122 | ||
| AK159272.1 | - | 6 - 7 | 990 | 122 | ||
| AK159731.1 | - | 6 - 7 | 990 | 122 | ||
| AK163933.1 | - | 6 - 7 | 1036 | 122 | ||
| BC138718.1 | - | 6 - 7 | 922 | 8861740 | 122 | |
| BC138723.1 | - | 6 - 7 | 922 | 8861745 | 122 | |
| J05287.1 | - | 6 - 7 | 962 | 122 | ||
| M32017.1 | - | 2 - 3 | 323 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lamp2
Genomic Map