Assay Details
| Gene Symbol: Lamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017959.2 | NP_001017959.1 | 7 - 8 | 1102 | 93 | |
| NM_001290485.1 | NP_001277414.1 | 7 - 8 | 1102 | 93 | ||
| NM_010685.4 | NP_034815.2 | 7 - 8 | 1102 | 93 | ||
| GenBank mRNA | AK012149.1 | - | 7 - 8 | 1061 | 93 | |
| AK032974.1 | - | 7 - 8 | 1064 | 93 | ||
| AK048174.1 | - | 7 - 8 | 1044 | 93 | ||
| AK078193.1 | - | 7 - 8 | 1096 | 93 | ||
| AK088804.1 | - | 8 - 9 | 1079 | 93 | ||
| AK128946.1 | - | 5 - 6 | 764 | 93 | ||
| AK136550.1 | - | 7 - 8 | 1063 | 93 | ||
| AK146731.1 | - | 7 - 8 | 1083 | 93 | ||
| AK150766.1 | - | 7 - 8 | 1071 | 93 | ||
| AK159272.1 | - | 7 - 8 | 1057 | 93 | ||
| AK159731.1 | - | 7 - 8 | 1057 | 93 | ||
| AK163933.1 | - | 7 - 8 | 1103 | 93 | ||
| BC138718.1 | - | 7 - 8 | 989 | 8861740 | 93 | |
| BC138723.1 | - | 7 - 8 | 989 | 8861745 | 93 | |
| J05287.1 | - | 7 - 8 | 1029 | 93 | ||
| M32017.1 | - | 3 - 4 | 390 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lamp2
Genomic Map