Assay Details
| Gene Symbol: Lamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290485.1 | NP_001277414.1 | 8 - 9 | 1266 | 100 | |
| GenBank mRNA | AK078193.1 | - | 8 - 9 | 1260 | 100 | |
More Information
Set Membership: |
|

Lamp2
Genomic Map