Assay Details
| Gene Symbol: Ngdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026890.2 | NP_081166.1 | 1 - 2 | 140 | 63 | |
| GenBank mRNA | BC052790.1 | - | 1 - 2 | 78 | 1515292 | 63 |
| CN715203.1 | - | 1 - 2 | 140 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC052790.1, NM_026890.2 transcripts.
Set Membership: |
|

Ngdn
Genomic Map