Assay Details
| Gene Symbol: Ngdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026890.2 | NP_081166.1 | 3 - 4 | 269 | 74 | |
| GenBank mRNA | AK005098.1 | - | 3 - 4 | 151 | 74 | |
| BC023770.1 | - | 2 - 3 | 154 | 74 | ||
| BC052790.1 | - | 3 - 4 | 207 | 1515292 | 74 | |
| EF437387.1 | - | 3 - 4 | 158 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ngdn
Genomic Map