Assay Details
| Gene Symbol: Ngdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026890.2 | NP_081166.1 | 5 - 6 | 494 | 69 | |
| GenBank mRNA | AK005098.1 | - | 5 - 6 | 376 | 69 | |
| AK212411.1 | - | 3 - 4 | 244 | 69 | ||
| AK214945.1 | - | 3 - 4 | 244 | 69 | ||
| BC023770.1 | - | 4 - 5 | 379 | 69 | ||
| BC052790.1 | - | 5 - 6 | 432 | 1515292 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ngdn
Genomic Map