Assay Details
| Gene Symbol: Lacc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172488.2 | NP_766076.1 | 4 - 5 | 1093 | 73 | |
| GenBank mRNA | AK033565.1 | - | 4 - 5 | 1094 | 73 | |
| AK053693.1 | - | 4 - 5 | 1378 | 73 | ||
| AK152154.1 | - | 4 - 5 | 1034 | 73 | ||
| AK153349.1 | - | 4 - 5 | 1118 | 73 | ||
| BC116748.1 | - | 3 - 4 | 849 | 40126067 | 73 | |
| BC119140.1 | - | 3 - 4 | 849 | 8733889 | 73 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Lacc1
Genomic Map