Assay Details
| Gene Symbol: N6amt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026526.2 | NP_080802.1 | 2 - 3 | 190 | 101 | |
| GenBank mRNA | AK005312.1 | - | 2 - 3 | 184 | 101 | |
| AK010911.1 | - | 1 - 2 | 185 | 101 | ||
| AK012083.1 | - | 2 - 3 | 191 | 101 | ||
| BC051925.1 | - | 2 - 3 | 181 | 5691329 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

N6amt2
Genomic Map