Assay Details
| Gene Symbol: Dhrs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037938.2 | NP_001033027.2 | 6 - 7 | 812 | 89 | |
| GenBank mRNA | AB045132.1 | - | 6 - 7 | 724 | 89 | |
| AK002691.1 | - | 2 - 3 | 253 | 89 | ||
| AK146988.1 | - | 6 - 7 | 692 | 89 | ||
| BC003484.1 | - | 6 - 7 | 647 | 2811468 | 89 | |
| BC054361.1 | - | 6 - 7 | 773 | 5008299 | 89 | |
| BQ032668.1 | - | 2 - 3 | 89 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dhrs4
Genomic Map