Assay Details
| Gene Symbol: Dhrs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037938.2 | NP_001033027.2 | 7 - 8 | 870 | 85 | |
| GenBank mRNA | AB045132.1 | - | 7 - 8 | 782 | 85 | |
| AK002691.1 | - | 3 - 4 | 311 | 85 | ||
| AK146988.1 | - | 7 - 8 | 750 | 85 | ||
| BC003484.1 | - | 7 - 8 | 705 | 2811468 | 85 | |
| BC054361.1 | - | 7 - 8 | 831 | 5008299 | 85 | |
| BQ032668.1 | - | 1 - 2 | 85 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dhrs4
Genomic Map