Assay Details
| Gene Symbol: Rec8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020002.3 | NP_064386.2 | 5 - 6 | 584 | 69 | |
| GenBank mRNA | AF262055.1 | - | 5 - 6 | 514 | 69 | |
| AK077167.1 | - | 5 - 6 | 519 | 69 | ||
| AK146832.1 | - | 5 - 6 | 570 | 69 | ||
| AU080225.1 | - | 5 - 6 | 585 | 69 | ||
| BC052155.1 | - | 5 - 6 | 580 | 6335959 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rec8
Genomic Map