Assay Details
| Gene Symbol: Dhrs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026819.3 | NP_081095.2 | 1 - 2 | 244 | 72 | |
| GenBank mRNA | AK003958.1 | - | 1 - 2 | 134 | 72 | |
| AK146317.1 | - | 1 - 2 | 191 | 72 | ||
| AK168115.1 | - | 1 - 2 | 191 | 72 | ||
| AK168147.1 | - | 1 - 2 | 191 | 72 | ||
| BC003930.1 | - | 1 - 2 | 175 | 3485584 | 72 | |
| BC023825.1 | - | 1 - 2 | 170 | 72 | ||
| BU963142.1 | - | 247 | 72 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003958.1, AK146317.1, AK168115.1, AK168147.1, BC003930.1, NM_026819.3 transcripts.
Set Membership: |
|

Dhrs1
Genomic Map