Assay Details
| Gene Symbol: Pebp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028526.4 | NP_082802.1 | 1 - 2 | 178 | 69 | |
| NM_028560.4 | NP_082836.2 | 1 - 2 | 178 | 69 | ||
| GenBank mRNA | AK006750.1 | - | 1 - 2 | 179 | 69 | |
| AK006964.1 | - | 1 - 2 | 169 | 69 | ||
| BC048621.1 | - | 1 - 2 | 156 | 6742909 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006750.1, AK006964.1, BC048621.1, NM_028526.4, NM_028560.4 transcripts.
Set Membership: |
|

Pebp4
Genomic Map