Assay Details
| Gene Symbol: Pebp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028560.4 | NP_082836.2 | 6 - 7 | 770 | 84 | |
| GenBank mRNA | CF104379.1 | - | 4 - 5 | 291 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pebp4
Genomic Map