Assay Details
| Gene Symbol: Trmt10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175389.4 | NP_780598.2 | 6 - 7 | 709 | 76 | |
| GenBank mRNA | AK089967.1 | - | 6 - 7 | 76 | ||
| AK146481.1 | - | 6 - 7 | 841 | 76 | ||
| BC022121.1 | - | 5 - 6 | 599 | 76 | ||
| BC090650.1 | - | 6 - 7 | 709 | 30651613 | 76 | |
| BC098478.1 | - | 6 - 7 | 774 | 30033713 | 76 | |
| BC141035.1 | - | 6 - 7 | 697 | 9055647 | 76 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Trmt10a
Genomic Map