Assay Details
| Gene Symbol: Bank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033350.2 | NP_001028522.2 | 14 - 15 | 2344 | 116 | |
| GenBank mRNA | AK041039.1 | - | 5 - 6 | 362 | 116 | |
| AK148665.1 | - | 3 - 4 | 139 | 116 | ||
| AY178734.1 | - | 14 - 15 | 2364 | 116 | ||
| BC116328.1 | - | 4 - 5 | 427 | 116 | ||
| EU051378.1 | - | 13 - 14 | 1945 | 116 | ||
More Information
Set Membership: |
|

Bank1
Genomic Map