Assay Details
| Gene Symbol: Gm29683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK049871.1 | - | 1 - 2 | 145 | 120 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930402F11 gene | 4930402F11Rik | -785.8kb | Anti-sense | Chr. 7 - 69496094 - 69499689 on NCBI Build GRCm38 | |
| Downstream |
nuclear receptor subfamily 2, group F, member 2 |
Nr2f2 |
2700033K02Rik, 9430015G03Rik, ARP-1, Aporp1, COUP-TF2, COUP-TFII, COUPTFB, EAR3, SVP40, Tcfcoup2 |
14.1kb | Sense | Chr. 7 - 70351946 - 70366746 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm29683
Genomic Map