Assay Details
| Gene Symbol: Chrna7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007390.3 | NP_031416.3 | 3 - 4 | 92 | ||
| GenBank mRNA | AF225980.1 | - | 3 - 4 | 284 | 92 | |
| AF325348.1 | - | 2 - 3 | 92 | |||
| AK034208.1 | - | 3 - 4 | 92 | |||
| AK157809.1 | - | 3 - 4 | 92 | |||
| AY574266.1 | - | 3 - 4 | 92 | |||
| AY851289.1 | - | 92 | ||||
| L37663.1 | - | 3 - 4 | 92 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Chrna7
Genomic Map