Assay Details
| Gene Symbol: Snrpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021336.4 | NP_067311.4 | 5 - 6 | 730 | 136 | |
| GenBank mRNA | AF230356.1 | - | 5 - 6 | 459 | 136 | |
| AK088438.1 | - | 5 - 6 | 505 | 136 | ||
| AK151800.1 | - | 5 - 6 | 523 | 136 | ||
| AK152546.1 | - | 5 - 6 | 523 | 136 | ||
| AK153502.1 | - | 5 - 6 | 523 | 136 | ||
| AK206837.1 | - | 4 - 5 | 293 | 136 | ||
| BC013777.1 | - | 5 - 6 | 497 | 2651704 | 136 | |
| BC029642.1 | - | 5 - 6 | 497 | 3588138 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snrpa1
Genomic Map