Assay Details
| Gene Symbol: Sytl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040085.2 | NP_001035174.1 | 13 - 14 | 2748 | 83 | |
| NM_001040087.2 | NP_001035176.1 | 4 - 5 | 955 | 83 | ||
| NM_001040088.2 | NP_001035177.1 | 4 - 5 | 883 | 83 | ||
| NM_001289583.1 | NP_001276512.1 | 14 - 15 | 2796 | 83 | ||
| NM_001289584.1 | NP_001276513.1 | 14 - 15 | 2715 | 83 | ||
| NM_001289585.1 | NP_001276514.1 | 5 - 6 | 1048 | 83 | ||
| NM_001289586.1 | NP_001276515.1 | 5 - 6 | 1003 | 83 | ||
| NM_031394.3 | NP_113571.2 | 13 - 14 | 2676 | 83 | ||
| GenBank mRNA | AB050742.1 | - | 4 - 5 | 494 | 83 | |
| AB057754.1 | - | 13 - 14 | 2135 | 83 | ||
| AB057755.1 | - | 7 - 8 | 2645 | 83 | ||
| AB057756.1 | - | 5 - 6 | 970 | 83 | ||
| AB057760.1 | - | 13 - 14 | 2054 | 83 | ||
| AB057761.1 | - | 12 - 13 | 2087 | 83 | ||
| AB057762.1 | - | 12 - 13 | 2015 | 83 | ||
| AI746961.1 | - | 4 - 5 | 428 | 83 | ||
| AK027924.1 | - | 4 - 5 | 866 | 83 | ||
| AK051653.1 | - | 5 - 6 | 1049 | 83 | ||
| AK173212.1 | - | 4 - 5 | 413 | 83 | ||
| BC060665.1 | - | 5 - 6 | 2498 | 83 | ||
| BC138714.1 | - | 12 - 13 | 1986 | 8861736 | 83 | |
| BC145398.1 | - | 13 - 14 | 2106 | 9053956 | 83 | |
| CA559030.1 | - | 3 - 4 | 389 | 83 | ||
| DV652808.1 | - | 7 - 8 | 700 | 83 | ||
More Information
Set Membership: |
|

Sytl2
Genomic Map