Assay Details
| Gene Symbol: Sytl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040085.2 | NP_001035174.1 | 14 - 15 | 2856 | 107 | |
| NM_001040087.2 | NP_001035176.1 | 5 - 6 | 1063 | 107 | ||
| NM_001040088.2 | NP_001035177.1 | 5 - 6 | 991 | 107 | ||
| NM_001289583.1 | NP_001276512.1 | 15 - 16 | 2904 | 107 | ||
| NM_001289584.1 | NP_001276513.1 | 15 - 16 | 2823 | 107 | ||
| NM_001289585.1 | NP_001276514.1 | 6 - 7 | 1156 | 107 | ||
| NM_001289586.1 | NP_001276515.1 | 6 - 7 | 1111 | 107 | ||
| NM_031394.3 | NP_113571.2 | 14 - 15 | 2784 | 107 | ||
| GenBank mRNA | AB050742.1 | - | 5 - 6 | 602 | 107 | |
| AB057754.1 | - | 14 - 15 | 2243 | 107 | ||
| AB057755.1 | - | 8 - 9 | 2753 | 107 | ||
| AB057756.1 | - | 6 - 7 | 1078 | 107 | ||
| AB057757.1 | - | 1 - 2 | 1069 | 107 | ||
| AB057760.1 | - | 14 - 15 | 2162 | 107 | ||
| AB057761.1 | - | 13 - 14 | 2195 | 107 | ||
| AB057762.1 | - | 13 - 14 | 2123 | 107 | ||
| AB057763.1 | - | 2 - 3 | 972 | 107 | ||
| AK027924.1 | - | 5 - 6 | 974 | 107 | ||
| AK051653.1 | - | 6 - 7 | 1157 | 107 | ||
| AK173212.1 | - | 5 - 6 | 521 | 107 | ||
| BC060665.1 | - | 6 - 7 | 2606 | 107 | ||
| BC138714.1 | - | 13 - 14 | 2094 | 8861736 | 107 | |
| BC145398.1 | - | 14 - 15 | 2214 | 9053956 | 107 | |
| CA559030.1 | - | 4 - 5 | 497 | 107 | ||
More Information
Set Membership: |
|

Sytl2
Genomic Map