Assay Details
| Gene Symbol: Lrrc28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175124.5 | NP_780333.3 | 8 - 9 | 993 | 76 | |
| NR_028143.1 | 8 - 9 | 898 | 76 | |||
| NR_028144.1 | 7 - 8 | 857 | 76 | |||
| GenBank mRNA | AK006352.1 | - | 2 - 3 | 199 | 76 | |
| AK077270.1 | - | 8 - 9 | 989 | 76 | ||
| AK159417.1 | - | 8 - 9 | 962 | 76 | ||
| BC027058.1 | - | 5 - 6 | 550 | 76 | ||
| BC052051.1 | - | 6 - 7 | 764 | 76 | ||
| BC086694.1 | - | 8 - 9 | 884 | 30697866 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc28
Genomic Map