Assay Details
| Gene Symbol: Pgpep1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030101.1 | NP_084377.1 | 2 - 3 | 208 | 71 | |
| GenBank mRNA | AK021221.1 | - | 2 - 3 | 209 | 71 | |
| BY728828.1 | - | 2 - 3 | 207 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK021221.1, NM_030101.1 transcripts.
Set Membership: |
|

Pgpep1l
Genomic Map