Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 137 | Mir137 |
Mirn137, mir-137, mmu-mir-137 |
-7.0 | Sense | Chr. 3 - 118433857 - 118433929 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 26871 |
Gm26871 | 45.6kb | Sense | Chr. 3 - 118504349 - 118528369 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map