Assay Details
| Gene Symbol: Mettl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201638.2 | NP_964000.2 | 4 - 5 | 488 | 128 | |
| GenBank mRNA | AK083169.1 | - | 4 - 5 | 430 | 128 | |
| AK129409.1 | - | 446 | 128 | |||
| AK146881.1 | - | 4 - 5 | 477 | 128 | ||
| AK153810.1 | - | 4 - 5 | 484 | 128 | ||
| BC052204.1 | - | 4 - 5 | 446 | 30007932 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl14
Genomic Map