Assay Details
| Gene Symbol: Mettl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201638.2 | NP_964000.2 | 5 - 6 | 573 | 123 | |
| GenBank mRNA | AK083169.1 | - | 5 - 6 | 515 | 123 | |
| AK089949.1 | - | 5 - 6 | 519 | 123 | ||
| AK129409.1 | - | 531 | 123 | |||
| AK146881.1 | - | 5 - 6 | 562 | 123 | ||
| AK153810.1 | - | 5 - 6 | 569 | 123 | ||
| BC052204.1 | - | 5 - 6 | 531 | 30007932 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl14
Genomic Map