Assay Details
| Gene Symbol: Mettl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201638.2 | NP_964000.2 | 9 - 10 | 1022 | 67 | |
| GenBank mRNA | AK146881.1 | - | 9 - 10 | 1011 | 67 | |
| AK153810.1 | - | 9 - 10 | 1018 | 67 | ||
| BC052204.1 | - | 9 - 10 | 980 | 30007932 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl14
Genomic Map