Assay Details
| Gene Symbol: Mettl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201638.2 | NP_964000.2 | 10 - 11 | 1223 | 156 | |
| GenBank mRNA | AK089949.1 | - | 10 - 11 | 1170 | 156 | |
| AK146881.1 | - | 10 - 11 | 1212 | 156 | ||
| BC052204.1 | - | 10 - 11 | 1181 | 30007932 | 156 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
nucleobase-containing compound metabolic process
stem cell population maintenance
methylation
mRNA destabilization
mRNA methylation
RNA binding
protein binding
methyltransferase activity
mRNA (2'-O-methyladenosine-N6-)-methyltransferase activity
transferase activity
mRNA splicing, via spliceosome
nucleobase-containing compound metabolic process
stem cell population maintenance
methylation
mRNA destabilization
mRNA methylation
RNA binding
protein binding
methyltransferase activity
mRNA (2'-O-methyladenosine-N6-)-methyltransferase activity
transferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22487801 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22487800 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22487802 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P377403 | AK089949.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P377403 | AK089949.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071566 | AK129409.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071566 | BC067020.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071566 | BC057981.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071566 | BC032094.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2071566 | BC038038.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919262 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 556073 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75953 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 64339 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923933 | AJ131957.2 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923933 | AK011298.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923933 | AK014138.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923933 | AK075930.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923933 | AK139556.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923933 | BC145789.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923933 | NM_133678.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 908175 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 780238 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375233 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945319 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 498864 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96753 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 866068 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919020 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 24134 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 33538 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 174194 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P101986 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P377403 | AK089949.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P377403 | AK089949.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22487781 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P101986 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05057115 | - | - |
| Whole Mouse Genome G4122A | A_52_P108186 | AK083169.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104155 | AK129409.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P377403 | AK089949.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P377403 | AK089949.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128296 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106366 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P101986 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03030735 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03030730 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03030732 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03030734 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22487800 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22487802 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22487801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22487795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03030730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02368748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03038293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487817 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487750 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02749142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02351776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487816 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03038294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03030732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03030734 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03050617 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02446338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02780869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02473783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22487812 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10501916 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501918 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501921 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501917 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501920 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501913 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501915 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501914 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501911 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501912 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501919 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454751_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10501909 | - | - |
| Mouse Genome 430 2.0 Array | 1454751_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454751_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 210529_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 210529_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300022753.mm.1 | NM_201638.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300022753.mm.1 | NM_201638.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300022753.mm.1 | TC0300002876.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300022754.mm.1 | NM_201638.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300022754.mm.1 | NM_201638.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300022754.mm.1 | TC0300002876.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1454751_at | - | - |
| Mouse Expression Set 430 | 1454751_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454751_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454751_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 931590 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2260021 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 107050026 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2260021 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3130427 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003130427 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002260021 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1224043 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1248621 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1224043 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1248621 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1238832 | - | - |
| mouse-6 v1.1 (Illumina) | scl0210529.1_20-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00210529.1_223-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2260021 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3130427 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 107050026 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0210529.1_20-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00210529.1_223-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2260021 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 107050026 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3130427 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2818206 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1248621 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1238832 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1248621 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2818206 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00210529.1_223-S | - | - |

Mettl14
Genomic Map