Assay Details
| Gene Symbol: Mettl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201638.2 | NP_964000.2 | 8 - 9 | 901 | 100 | |
| GenBank mRNA | AK129409.1 | - | 859 | 100 | ||
| AK146881.1 | - | 8 - 9 | 890 | 100 | ||
| AK153810.1 | - | 8 - 9 | 897 | 100 | ||
| BC052204.1 | - | 8 - 9 | 859 | 30007932 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl14
Genomic Map