Assay Details
| Gene Symbol: Ap1ar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145964.2 | NP_666076.2 | 1 - 2 | 339 | 70 | |
| GenBank mRNA | CJ169738.1 | - | 1 - 2 | 340 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ap1ar
Genomic Map