Assay Details
| Gene Symbol: Ap1ar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145964.2 | NP_666076.2 | 2 - 3 | 397 | 85 | |
| GenBank mRNA | AK200436.1 | - | 59 | 85 | ||
| BB611022.1 | - | 2 - 3 | 85 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ap1ar
Genomic Map