Assay Details
| Gene Symbol: Dpyd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170778.2 | NP_740748.1 | 2 - 3 | 247 | 107 | |
| GenBank mRNA | AK149426.1 | - | 2 - 3 | 249 | 107 | |
| BC039699.1 | - | 2 - 3 | 248 | 4235971 | 107 | |
More Information
Set Membership: |
|

Dpyd
Genomic Map