Assay Details
| Gene Symbol: Tenm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011855.4 | NP_035985.2 | 20 - 21 | 3533 | 94 | |
| GenBank mRNA | AB025410.1 | - | 20 - 21 | 3535 | 94 | |
| BC138860.1 | - | 19 - 20 | 3516 | 8861882 | 94 | |
| BC138861.1 | - | 19 - 20 | 3516 | 8861883 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tenm1
Genomic Map