Assay Details
| Gene Symbol: Tenm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011855.4 | NP_035985.2 | 28 - 29 | 5481 | 84 | |
| GenBank mRNA | AB025410.1 | - | 28 - 29 | 5483 | 84 | |
| AK037897.1 | - | 1 - 2 | 170 | 84 | ||
| BC138860.1 | - | 27 - 28 | 5464 | 8861882 | 84 | |
| BC138861.1 | - | 27 - 28 | 5464 | 8861883 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tenm1
Genomic Map