Assay Details
| Gene Symbol: Tenm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011855.4 | NP_035985.2 | 4 - 5 | 630 | 72 | |
| GenBank mRNA | AB025410.1 | - | 4 - 5 | 632 | 72 | |
| AK034286.1 | - | 6 - 7 | 1523 | 72 | ||
| AK047423.1 | - | 6 - 7 | 1398 | 72 | ||
| BC138860.1 | - | 3 - 4 | 613 | 8861882 | 72 | |
| BC138861.1 | - | 3 - 4 | 613 | 8861883 | 72 | |
More Information
Set Membership: |
|

Tenm1
Genomic Map