Assay Details
| Gene Symbol: Tnni3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177066.5 | NP_796040.3 | 10 - 11 | 1121 | 66 | |
| GenBank mRNA | AY526095.1 | - | 10 - 11 | 1121 | 66 | |
| AY526096.1 | - | 10 - 11 | 1121 | 66 | ||
| BC139368.1 | - | 10 - 11 | 1054 | 8862390 | 66 | |
| BC139394.1 | - | 10 - 11 | 1054 | 8862416 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tnni3k
Genomic Map