Assay Details
| Gene Symbol: Setdb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081024.1 | NP_001074493.1 | 8 - 9 | 1632 | 69 | |
| GenBank mRNA | BC157926.1 | - | 7 - 8 | 1454 | 9088099 | 69 |
| BC172021.1 | - | 7 - 8 | 1454 | 9054307 | 69 | |
| EU155090.1 | - | 7 - 8 | 1446 | 69 | ||
| EU155091.1 | - | 7 - 8 | 1446 | 69 | ||
| EU155093.1 | - | 6 - 7 | 1380 | 69 | ||
| EU155094.1 | - | 6 - 7 | 1380 | 69 | ||
| EU155096.1 | - | 6 - 7 | 1103 | 69 | ||
| EU155097.1 | - | 8 - 9 | 1505 | 69 | ||
| EU155098.1 | - | 6 - 7 | 1398 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Setdb2
Genomic Map