Assay Details
| Gene Symbol: 1700003H04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005639.1 | - | 5 - 6 | 584 | 71 | |
| BC100480.1 | - | 5 - 6 | 518 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700006A11 gene | 1700006A11Rik | -147.2kb | Sense | Chr. 3 - 124400984 - 124426028 on NCBI Build GRCm38 | |
| Downstream | N-deacetylase/N-sulfotransferase (heparin glucosaminyl) 4 | Ndst4 | 4930439H17Rik | 829.5kb | Anti-sense | Chr. 3 - 125404076 - 125728351 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

1700003H04Rik
Genomic Map