Assay Details
| Gene Symbol: Lins | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191001.1 | NP_001177930.1 | 6 - 7 | 1601 | 108 | |
| NM_152815.2 | NP_690028.2 | 6 - 7 | 1616 | 108 | ||
| NR_034030.1 | 6 - 7 | 1530 | 108 | |||
| GenBank mRNA | AB083158.1 | - | 4 - 5 | 1200 | 108 | |
| AK012554.1 | - | 6 - 7 | 1463 | 108 | ||
| AK156066.1 | - | 6 - 7 | 1585 | 108 | ||
| AK171170.1 | - | 6 - 7 | 1528 | 108 | ||
| BC030943.1 | - | 8 - 9 | 1608 | 108 | ||
| BC058648.1 | - | 6 - 7 | 1433 | 108 | ||
| BC059228.1 | - | 1 - 2 | 602 | 108 | ||
| BC138549.1 | - | 4 - 5 | 1274 | 8861569 | 108 | |
More Information
Set Membership: |
|

Lins
Genomic Map