Assay Details
| Gene Symbol: Esrrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243792.1 | NP_001230721.1 | 1 - 2 | 205 | 117 | |
| GenBank mRNA | AK052731.1 | - | 1 - 2 | 184 | 117 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK052731.1, NM_001243792.1 transcripts.
Set Membership: |
|

Esrrg
Genomic Map