Assay Details
| Gene Symbol: Qk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159516.1 | NP_001152988.1 | 3 - 4 | 896 | 111 | |
| NM_001159517.1 | NP_001152989.1 | 3 - 4 | 896 | 111 | ||
| NM_021881.2 | NP_068681.1 | 3 - 4 | 896 | 111 | ||
| GenBank mRNA | AF090402.1 | - | 4 - 5 | 1056 | 111 | |
| AF090403.1 | - | 4 - 5 | 1056 | 111 | ||
| AF091047.1 | - | 1056 | 111 | |||
| AK005169.1 | - | 3 - 4 | 282 | 111 | ||
| AK132303.1 | - | 3 - 4 | 897 | 111 | ||
| BC053426.1 | - | 4 - 5 | 568 | 6335004 | 111 | |
| BC056346.1 | - | 3 - 4 | 422 | 5705171 | 111 | |
| U44940.1 | - | 3 - 4 | 915 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Qk
Genomic Map