Assay Details
| Gene Symbol: Nupl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006062.1 | - | 14 - 15 | 1804 | 85 | |
| AK078279.1 | - | 14 - 15 | 1749 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nupl1| Gene Symbol: Nupl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006062.1 | - | 14 - 15 | 1804 | 85 | |
| AK078279.1 | - | 14 - 15 | 1749 | 85 | ||
Set Membership: |
|