Assay Details
| Gene Symbol: Pebp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028560.4 | NP_082836.2 | 3 - 4 | 511 | 71 | |
| GenBank mRNA | AK006964.1 | - | 3 - 4 | 502 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pebp4
Genomic Map