| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200000261.mm.1 |
NM_001278581.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000261.mm.1 |
NM_001278582.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000261.mm.1 |
NM_001278583.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000261.mm.1 |
NM_001278584.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000261.mm.1 |
NM_008895.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000261.mm.1 |
TC1200000017.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200000265.mm.1 |
NM_001278581.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000265.mm.1 |
NM_001278582.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000265.mm.1 |
NM_001278583.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000265.mm.1 |
NM_001278584.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000265.mm.1 |
NM_008895.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000265.mm.1 |
TC1200000017.mm.1 |
MTA TC |