Assay Details
| Gene Symbol: Mtrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145960.4 | NP_666072.3 | 8 - 9 | 1369 | 61 | |
| GenBank mRNA | AK043981.1 | - | 8 - 9 | 1175 | 61 | |
| AK051834.1 | - | 8 - 9 | 1155 | 61 | ||
| AK077749.1 | - | 8 - 9 | 1370 | 61 | ||
| BC028898.1 | - | 8 - 9 | 1285 | 4205917 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtrf1
Genomic Map