Assay Details
| Gene Symbol: Stpg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC138108.1 | - | 2 - 3 | 261 | 8861127 | 76 |
| BC138109.1 | - | 2 - 3 | 261 | 8861128 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stpg2| Gene Symbol: Stpg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC138108.1 | - | 2 - 3 | 261 | 8861127 | 76 |
| BC138109.1 | - | 2 - 3 | 261 | 8861128 | 76 | |
Set Membership: |
|