Assay Details
| Gene Symbol: Gm28505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK080902.1 | - | 1 - 2 | 154 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
Parkinson disease (autosomal recessive, juvenile) 2, parkin |
Park2 | PRKN | Anti-sense | Chr. 17 - 10840384 - 12063370 on NCBI Build GRCm38 | |
| Downstream |
DNA segment, Chr 17, ERATO Doi 648, expressed |
D17Ertd648e | 773.6kb | Sense | Chr. 17 - 11863070 - 11883189 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm28505
Genomic Map