Assay Details
| Gene Symbol: A730004F24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042551.1 | - | 2 - 3 | 499 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lysophospholipase-like 1 | Lyplal1 |
BC027340, Q96AVO |
-394.6kb | Sense | Chr. 1 - 186087732 - 186117310 on NCBI Build GRCm38 |
| Downstream |
transforming growth factor, beta 2 |
Tgfb2 |
BB105277, Tgf-beta2, Tgfb-2 |
103.5kb | Sense | Chr. 1 - 186622787 - 186709033 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 517474 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 413025 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 208055 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 167511 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 780188 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666111 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 761776 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75348 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 73976 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108242 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 391561 | AK016019.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913272 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 58568 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955389 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 175224 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 51014 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106245 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04388473 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106245 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04388516 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04388491 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112962 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106245 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04388473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00245383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00859103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907510 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907521 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00670767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00363246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907725 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00416008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00388279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00605362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00526298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907604 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907724 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907570 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907738 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907454 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907643 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00375038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907536 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00388274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907743 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907448 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00570827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00409184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00819962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907737 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00142072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907600 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00183009 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00859106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00605370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00975113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00582390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00582328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907440 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00605368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00391245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907467 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907733 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00255008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907579 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00245384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00295213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907526 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907572 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00819970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00605363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907442 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907544 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00017685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00043496 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00183022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00859108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907444 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907519 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907637 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00605358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00858332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907565 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00388275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00859088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00414511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907575 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00858339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00605371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907361 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00137706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907727 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00670768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907545 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00363243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907750 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907517 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20907425 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103060113 | - | - |

A730004F24Rik
Genomic Map