Assay Details
| Gene Symbol: 6530411M01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027881.1 | 1 - 2 | 740 | 66 | ||
| GenBank mRNA | AK078247.1 | - | 1 - 2 | 555 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700010I14 gene | 1700010I14Rik | -140.8kb | Anti-sense | Chr. 17 - 8988320 - 9008319 on NCBI Build GRCm38 | |
| Downstream |
poly(A) binding protein, cytoplasmic 6 |
Pabpc6 |
4932702K14Rik, AI428050, Pabpc3 |
498.5kb | Sense | Chr. 17 - 9666497 - 9669704 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

6530411M01Rik
Genomic Map