Assay Details
| Gene Symbol: Gm32693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039352.1 | - | 2 - 3 | 267 | 136 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RAP1, GTP-GDP dissociation stimulator 1 |
Rap1gds1 |
BC011279, GDS1 |
-702.0kb | Sense | Chr. 3 - 138925897 - 139075201 on NCBI Build GRCm38 |
| Downstream | microRNA 7j | Mir7j |
Gm27716, Mirlet7j, mmu-let-7j |
247.8kb | Anti-sense | Chr. 3 - 140028249 - 140028370 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm32693
Genomic Map